Documenti fondamentali
Z154075
Sigma-Aldrich
Pinch clamps for SJ joints
joint: SJ 35/20
Scegli un formato
1 EA
CHF 50.20
CHF 50.20
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 50.20
About This Item
Codice UNSPSC:
41122413
NACRES:
NB.35
CHF 50.20
Check Cart for Availability
Materiali
stainless steel
Giunto
joint: SJ35/20
Categorie correlate
Descrizione generale
For use with Z137294, and for Ace pressure filter/drying funnels.
Use with flash-chromatography ball jointed assemblies and rotary evaporator receiver flasks.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 7
1 of 7
Labeling tape W × L 3/4 in. × 500 in., red
Z768332
Labeling tape
Labeling tape blue, W × L 3/4 in. × 500 in.
Z768324
Labeling tape
Labeling tape green, W × L 3/4 in. × 500 in.
Z768316
Labeling tape
Labeling tape yellow, W × L 3/4 in. × 500 in.
Z768308
Labeling tape
Labeling tape white, W × L 3/4 in. × 500 in.
Z768294
Labeling tape
Zinc chloride puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥98%
Sigma-Aldrich
31650
Zinc chloride