Documenti
THSD1447
ThalesNano MidiCart catalyst cartridge system
L × diam. 116 mm × 9.2 mm, nickel sponge
Sinonimo/i:
Heterogeneous catalysis system, safe Heterogeneous catalysis
About This Item
Codice UNSPSC:
41103817
NACRES:
NB.51
Produttore/marchio commerciale
ThalesNano THS D1447
L × diam.
116 mm × 9.2 mm
Descrizione generale
MiDiCarts are suitable for use only with Thalesnano H-Cube MidiCart reactors.
Note legali
MidiCart is a trademark of ThalesNano Nanotechnology Inc.