Key Documents
SYNC291910
Synthware graduated concentrator tube
10 mL, joint: ST/NS 19/22
Sinonimo/i:
Synthware C291910, glass concentrator tube
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.51
Materiali
colorless tube
glass tube
Giunto
joint: ST/NS19/22
agenzia
suitable for EPA 1613
Sterilità
non-sterile
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware C291910
Capacità
10 mL
Descrizione generale
Graduated concentrator tubes for use with Kuderna-Danish evaporative set-ups. Tube can be stoppered for storage. Overall height is 100 mm for 10 mL, and 170 mm for the 25 mL.
Note legali
Synthware is a trademark of Kemtech America Inc.
Visita l’Archivio dei documenti
Contenuto correlato
This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.