Documenti fondamentali
SYNB262924
Synthware bubbler with PTFE stopcock
top joint: ST/NS 29/42, bottom joint: ST/NS 24/40
Sinonimo/i:
Synthware B262924, mineral oil bubbler
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.51
Giunto
bottom joint: ST/NS24/40
joint: ST/NS top29/42
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware B262924
Descrizione generale
With small ball joints to prevent mineral oil being sucked back under sudden negative pressure in the system.
Note legali
Synthware is a trademark of Kemtech America Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)