Documenti fondamentali
S80404
2-CHLORO-5-NITRO-BENZOIC ACID METHYL ESTER
AldrichCPR
Scegli un formato
S80404-250MG
CHF 132.00
CHF 132.00
Spedizione prevista il24 maggio 2025
Scegli un formato
Cambia visualizzazione
S80404-250MG
CHF 132.00
About This Item
Formula condensata:
C8H6ClNO4
Numero CAS:
Peso molecolare:
215.594
Numero MDL:
CHF 132.00
Spedizione prevista il24 maggio 2025
Altre note
Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of rare and unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH'S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT AS-IS AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)