Documenti fondamentali
S240737
2,4,6-TRIMETHYL-1,4-DIHYDRO-PYRIDINE-3,5-DICARBOXYLIC ACID DIETHYL ESTER
AldrichCPR
Scegli un formato
S240737-25MG
CHF 174.00
CHF 174.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
S240737-25MG
CHF 174.00
About This Item
Formula condensata:
C14H21NO4
Numero CAS:
Peso molecolare:
267.328
Numero MDL:
CHF 174.00
Check Cart for Availability
Altre note
Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of rare and unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH'S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT AS-IS AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)