Documenti fondamentali
CLS426012L
Pyrex® round-bottom boiling flask, short ring neck
capacity 12,000 mL
Sinonimo/i:
bottom flask, flask round, round flask, Corning® 4260
About This Item
Codice UNSPSC:
41121804
NACRES:
NB.43
Prezzi e disponibilità al momento non sono disponibili
Materiali
borosilicate glass
round bottom flask
short
Misura tappo a cono
11
Sterilità
non-sterile
Caratteristiche
closure type Standard Taper joint
Confezionamento
case of 1
Produttore/marchio commerciale
Corning 4260-12L
Capacità
12,000 mL
Dimens. collo
(1)
Descrizione generale
These round bottom boiling flasks have short ring necks and are designed to be used with rubber stoppers. They are precisely tooled for uniform stopper fit. The reinforced rim increases mechanical strength. These flasks are used in applications requiring rigid stoppering and as wash bottles in routine lab washing. Wall thicknesses are carefully controlled to provide the proper balance between thermal expansion and mechanical strength for greater shock resistance. Their excellent chemical durability results in contamination-free results and long life under the toughest lab conditions.
Note legali
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)