Documenti fondamentali
CAD4019
Standard point hypodermic needles, 304 stainless steel
gauge 25, L 5/8 in., nickel plated hub
Scegli un formato
12 EA
CHF 184.00
CHF 184.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
12 EA
CHF 184.00
About This Item
Codice UNSPSC:
42142523
NACRES:
NB.03
CHF 184.00
Check Cart for Availability
Materiali
304 stainless steel needle
nickel plated hub
Confezionamento
pack of 12 ea
Produttore/marchio commerciale
Cadence Science 4019
Lunghezza
5/8 in.
Gauge
25
Categorie correlate
Descrizione generale
Hypodermic needles are precision made and designed to ensure strict adherence with U.S. Government specification GG-N-196. Points are machine ground to achieve uniform sharpness. Precision grinding, micro-bead jet blasting, and ultrasonic cleaning assure freedom from burrs and debris. All needle cannula are fabricated of type 304 stainless steel tubing which must pass rigorous quality control prior to acceptance and use.
Standard nickle plated Luer hubs comply with Luer taper specifications ISO 594. This assures an interchangeable fit mating with Luer slips and Luer locks. Contoured hub cavity facilitates cleaning.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)