Documenti fondamentali
BR782840
BRAND® insert strips for 24 x 6-well BRANDplates®
PC membrane, pore size 3 μm
Sinonimo/i:
BRANDplates®, Cell culture, Insert Strips, Insert System, Inserts, Tissue culture
Scegli un formato
12 EA
CHF 292.00
CHF 292.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
12 EA
CHF 292.00
About This Item
Codice UNSPSC:
41122107
NACRES:
NB.22
CHF 292.00
Check Cart for Availability
Materiali
PC membrane
Descrizione
cellGrade plus
Sterilità
sterile
Confezionamento
pack of 12 ea (strips of 4 inserts, individually wrapped)
Produttore/marchio commerciale
BRAND 782840
Misura
6 well
Dimensione pori
3 μm
Categorie correlate
Descrizione generale
Innovative cell culture plates and inserts for manual and automatic cell and tissue culture. The BRANDplates® Insert System, developed in conjunction with the Fraunhofer company, offers an innovative expansion to BRAND′s existing cell culture product range. It consists of two different cell culture plates and two designs of cell culture inserts that can be combined in different configurations.Insert specification:4x insert strips, can be divided into up to 4 individual inserts - hanging position in wellHanging inserts – distance between the well base and insert membrane is 4 mmIndividually packaged strips inserted in thermoformed beakers with 6 strips each in the 6-well cell culture platecellGrade plus (hydrophilic cell culture-) surface.Smooth wall inserts: For standard applications like secretion studies, co-cultivations, chemotaxis tests, etc.PS cellGrade plus surface, sterile, strip of 4 inserts (divisible).BIO-CERT® CELL CULTURE QUALITY: Free of human DNA, RNase, DNase, Pyrogens and non cytotoxic
Note legali
BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
BRANDplates is a registered trademark of BRAND GMBH + CO KG
cellGrade is a trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)