Documenti fondamentali
BR781500
Brand®1.2 ML Tubes
with 96 individual tubes, polypropylene
Sinonimo/i:
Sample Storage, Tube racks, Tubes
Scegli un formato
1 PKG
CHF 122.00
CHF 122.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 PKG
CHF 122.00
About This Item
Codice UNSPSC:
41122804
NACRES:
NB.22
CHF 122.00
Check Cart for Availability
Materiali
polypropylene
Sterilità
non-sterile
Caratteristiche
with 96 individual tubes
Produttore/marchio commerciale
BRAND 781500
Confezionamento
pkg of (10 racks of 96 individual tubes)
Capacità
1.2 mL
Descrizione generale
PP. Tubes and rack are autoclavable at 121 °C (2 bar), acc. DIN EN 285. Ideal for biological tests, such as PCR, cell uptake studies, RIA and EIA. Designed for storing and freezing down to -80 °C, and for transporting reagents and samples. Compatible with standard microplates and suitable for working with multichannel pipettes. (Size of tubes: outer Ø 8.8 mm, height 45 mm.) Single caps and strip caps, PE, available as accessories.BIO-CERT® CERTIFIED QUALITY: Free of human DNA, RNase, DNase and Pyrogens
Note legali
BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)