BR701372
BRAND® sealing film for microplates
self-adhesive, suitable for (for luminescence measurement), vinyl
Sinonimo/i:
Accessories, PCR, Plates, Sealing films
About This Item
Codice UNSPSC:
41122410
NACRES:
NB.22
Prodotti consigliati
Materiali
vinyl
vinyl film
white
Sterilità
non-sterile
Caratteristiche
self-adhesive
Confezionamento
pack of 50 ea
Produttore/marchio commerciale
BRAND 701372
Compatibilità
suitable for (for luminescence measurement)
Descrizione generale
White and black films provide support for fluorescence and luminescence measurements and for microscopy. Weak signals are amplified and interference of light from outside is minimized.
Caratteristiche e vantaggi
- Black, light-absorbing film for optimal fluorescence measurements
- White, light-reflecting film for amplified luminescent signal
- Storable down to -40 °C
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)