Documenti fondamentali
BR701370
BRAND® sealing film for microplates
self-adhesive, suitable for (for automation), polypropylene
Sinonimo/i:
Accessories, PCR, Plates, Sealing films
Scegli un formato
50 EA
CHF 184.00
CHF 184.00
Spedizione prevista il01 giugno 2025
Scegli un formato
Cambia visualizzazione
50 EA
CHF 184.00
About This Item
Codice UNSPSC:
41122410
NACRES:
NB.22
CHF 184.00
Spedizione prevista il01 giugno 2025
Materiali
film (PP/PE-HD)
polypropylene
Sterilità
non-sterile
Caratteristiche
piercable
self-adhesive
Confezionamento
pack of 50 ea
Produttore/marchio commerciale
BRAND 701370
Compatibilità
suitable for (for automation)
Descrizione generale
Due to the increasing number of analysis methods, the number of samples processed is growing steadily. This means that high-throughput analyses are gaining significance. In these automated processes, the films must not only protect the samples, but also comply with the mechanical requirements of automation.
Caratteristiche e vantaggi
  • Pierceability
  • Adhesive-free zones for contamination-free tasks
  • Tight seal for minimizing evaporation
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)