Documenti fondamentali
BR476900
BRAND® slide box
Holds 5 or 10 x slides 76x26 mm
Sinonimo/i:
Microscope slides, Microscopy, Slide box, Staining, Staining Accessories
Scegli un formato
10 EA
CHF 74.70
CHF 74.70
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
10 EA
CHF 74.70
About This Item
Codice UNSPSC:
41123302
NACRES:
MA.01
CHF 74.70
Check Cart for Availability
Materiali
polypropylene
Caratteristiche
Holds 5 or 10 x slides 76x26 mm
Confezionamento
pack of 10 ea
Produttore/marchio commerciale
BRAND 476900
Descrizione generale
Sturdy design. Screw lid with dealing wire lug. Ideal for storing or shipping 5 thick or 10 thin slides. Slides protrude 10 mm above the box, to facilitate removal.
For slides, size (in, mm) 76 x 26. Numbered slots in base. Lid with index card.
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Jiasheng Wang et al.
Bioscience, biotechnology, and biochemistry, 84(11), 2215-2227 (2020-07-24)
This study explored the mechanism of NEAT1 in sepsis-induced AKI rats. Cecal ligation punctures (CLP)-induced AKI rats were injected with siRNA-NEAT1 lentivirus. Kidney histopathology and apoptosis were evaluated via hematoxylin-eosin and TUNEL staining, respectively. ELISA determined the levels of Blood