Documenti fondamentali
BR4600240
BRAND® Dispensette® S Fixed-volume bottle-top dispenser
volume 10 mL, without recirculation valve
Scegli un formato
1 EA
CHF 360.00
CHF 360.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 360.00
About This Item
Codice UNSPSC:
41121500
NACRES:
NB.01
CHF 360.00
Check Cart for Availability
Caratteristiche
DE-M marking
without recirculation valve
Produttore/marchio commerciale
BRAND 4600240
Volume
10 mL
Raccordi
(GL 28/S28)
GL 32
GL 38
S 40
Descrizione generale
Dispensette S is suitable for an amazingly broad range of applications, particularly for many acids, saline solutions and organic solvents.
Caratteristiche e vantaggi
  • Extremely easy filling and dispensing
  • Simple priming by optimized flow channels
  • Rapid and safe volume adjustment with internal scalloped track
Altre note
Fixed Volume- For standard applications. Simple calibration adjustment with a supplied tool.
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Contenuto correlato
Chemical compatibility chart for all Dispensette® S and Dispensette® S Organic models.