Documenti fondamentali
BR41620
BRAND® graduated cylinder low form embossed scale
volume 25 mL, polypropylene
Sinonimo/i:
Graduated Cylinders, Plastics
Scegli un formato
10 EA
CHF 29.40
CHF 29.40
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
10 EA
CHF 29.40
About This Item
Codice UNSPSC:
41121805
NACRES:
NB.43
CHF 29.40
Check Cart for Availability
Materiali
polypropylene
Produttore/marchio commerciale
BRAND 41620
Volume
25 mL
Descrizione generale
The BRAND® Graduated Cylinder, Low Form with Embossed Scale, is designed for precise liquid measurement in laboratory settings. Made from high-clarity polypropylene (PP), this cylinder is calibrated to contain (TC, In), ensuring accurate volume readings. Its low form design enhances stability and ease of use, while the embossed scale provides clear visibility for quick reference. This graduated cylinder can withstand exposure to temperatures up to 80 °C without exceeding permanent error limits, making it suitable for a variety of applications. Reliable and durable, this graduated cylinder is an essential tool for any laboratory.
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)