Documenti fondamentali
BR115016
BRAND® sample vial
for Technicon-Anal., capacity 2.0 mL
Sinonimo/i:
Sample Cups, Sample vials
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.11
Prezzi e disponibilità al momento non sono disponibili
Materiali
colorless tube
polystyrene
polystyrene tube
self-standing bottom
Sterilità
non-sterile
Caratteristiche
graduations: no
Confezionamento
pack of 1000 ea
Produttore/marchio commerciale
BRAND 115016
Capacità
2.0 mL
Descrizione generale
The BRAND® Sample Vial is designed for secure and efficient storage of samples in laboratory settings. Made from high-quality transparent polystyrene (PS), this vial allows for easy visibility of contents, facilitating quick identification and monitoring.
Caratteristiche e vantaggi
  • BRAND® sample vial is designed for various laboratory applications.
  • Made of transparent polystyrene for accurate and quick sample analysis.
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)