Documenti fondamentali
BAH186100470
Stak-A-Tray small tray
W × L 17.8 cm × 35.6 cm
Scegli un formato
1 EA
CHF 113.00
CHF 113.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 113.00
About This Item
Codice UNSPSC:
41122800
NACRES:
NB.35
CHF 113.00
Check Cart for Availability
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Bel-Art H18610-0470
Largh. × lungh.
17.8 cm × 35.6 cm
Descrizione generale
Modular rack and tray system allows multi-layer storage and prevents tipping of stacked flasks that result in spills.
Caratteristiche e vantaggi
  • Can be used with Petri dishes and cell culture flasks in incubators, cold rooms or on the bench
  • Each tier accommodates one large or two small trays
  • Stack up to five racks for increased storage within the same footprint
  • Perforated trays allow air flow and slide easily in the rack
  • All parts are stainless steel; autoclavable at 121ºC (250ºF)
  • Racks and trays sold separately
Note legali
Stak-A-Tray is a trademark of SP Scienceware, a division of SP
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)