Vai a
HS2344A
Reversible PCR Tube Rack
to hold, 168 x 0.2 mL/ 40 x 0.5 mL/12 x 1.5-2.0 mL tubes, assorted colors polypropylene
Autenticati per visualizzare i prezzi organizzativi e contrattuali.
Scegli un formato
Cambia visualizzazione
A voi/SKUDisponibilitàPrezzo
5 ea
Per conoscere la disponibilità, visualizza il carrello
CHF 71.70
Informazioni su questo articolo
NACRES:
NB.12
UNSPSC Code:
41122800
Manufacturer/tradename:
Heathrow Scientific HS2344A
Feature:
to hold 168 x 0.2 mL/ 40 x 0.5 mL/12 x 1.5-2.0 mL tubes
Material:
assorted colors polypropylene
Packaging:
pack of 5 ea
CHF 71.70
Per conoscere la disponibilità, visualizza il carrello
Servizio Tecnico
Hai bisogno di aiuto? Il nostro team di scienziati qualificati è a tua disposizione.
Permettici di aiutarti
material
assorted colors polypropylene
feature
to hold 168 x 0.2 mL/ 40 x 0.5 mL/12 x 1.5-2.0 mL tubes
packaging
pack of 5 ea
manufacturer/tradename
Heathrow Scientific HS2344A
L × W × H
206 mm × 131 mm × 54 mm
General description
The PCR Racks are reversible and allow you to use 0.2,0.5, and 1.5/2.0mL tubes. The removable hinged lid fits both sides of rack.
Confronta articoli simili
Visualizza il confronto per intero
Mostra le differenze
1 of 1
Questo articolo
Z692891R6651R6401
Reversible PCR Tube Rack to hold, 168 x 0.2 mL/ 40 x 0.5 mL/12 x 1.5-2.0 mL tubes, assorted colors polypropylene
HS2344A
Reversible PCR Tube Rack
Rota-Rack® Duo tube rack pkg of 1 ea
Z692891
Rota-Rack® Duo tube rack
Reversible 96 well rack pink
R6651
Reversible 96 well rack
Reversible 96 well rack blue
R6401
Reversible 96 well rack
description
to hold, 168 x 0.2 mL/ 40 x 0.5 mL/12 x 1.5-2.0 mL tubes, assorted colors polypropylene
description
pkg of 1 ea
description
pink
description
blue
feature
to hold 168 x 0.2 mL/ 40 x 0.5 mL/12 x 1.5-2.0 mL tubes
feature
Holds 48 x 0.5 mL tubes, Holds 48 x 1.5 mL tubes, Holds 96 x 0.2 mL tubes
feature
Holds 96 x 0.5-2 mL tubes
feature
Holds 96 x 0.5-2 mL tubes
manufacturer/tradename
Heathrow Scientific HS2344A
manufacturer/tradename
Heathrow Scientific HS29041A
manufacturer/tradename
Heathrow Scientific HS2345E
manufacturer/tradename
Heathrow Scientific HS2345C
material
assorted colors polypropylene
material
polypropylene
material
polypropylene
material
polypropylene
packaging
pack of 5 ea
packaging
pkg of 1 ea
packaging
-
packaging
-
L × W × H
206 mm × 131 mm × 54 mm
L × W × H
296 mm × 118 mm × 34 mm
L × W × H
24.4 cm (9.6 in.) × 12 cm (4.7 in.) × 5.1 cm (2 in.)
L × W × H
24.4 cm (9.6 in.) × 12 cm (4.7 in.) × 5.1 cm (2 in.)
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)