Z635146
Sartorius mLINE® mechanical pipettor
Single channel, volume 2-20 μL
Synonyme(s) :
Biohit pipettes, pipettor
414.00 CHF
About This Item
414.00 CHF
Produits recommandés
Caractéristiques
electronic: no
single-channel
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Sartorius 725030
Plage de mesure du volume
2-20 μL
Volume
20 μL
Catégories apparentées
Description générale
- Extremely light pipetting and tip ejection
- Unique safety features
- Stepless volume adjustment and patented volume locking system
- Thermal insulation of internal components to improve accuracy
- Full autoclavability without disassembling
- Easy cleaning, maintenance and calibration
- ID tag and color-coded caps
- Safe-Cone Filters and unique ejection
- Opti-load tip mounting mechanism for even tip sealing and light tip ejection in multichannel pipettors