Principaux documents
TMO249720
Nunc® breathable cell culture sealing film
rayon acrylate, non-sterile, case of 50 ea
Synonyme(s) :
adhesive film, adhesive foil
50 EACH
172.00 CHF
172.00 CHF
Date d'expédition estimée le01 juin 2025
Changer de vue
50 EACH
172.00 CHF
About This Item
Code UNSPSC :
41161502
Nomenclature NACRES :
NB.15
172.00 CHF
Date d'expédition estimée le01 juin 2025
Stérilité
non-sterile
Conditionnement
case of 50 ea
Fabricant/nom de marque
Nunc 249720
Catégories apparentées
Description générale
Minimize evaporation and protect samples from contamination and spilling with Thermo Scientific Nunc Sealing Tapes. These adhesive seals are ideal for all assay applications. Nunc Life Science products offer reliability and reproducible results, and are backed by industry-leading quality standards and certifications.
Informations légales
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries