Documents
IST108078TS
ThermASeal Foil heat sealable seal
LabSheet, W × L 78 mm × 125 mm, pkg of pack of 5 sheets (trial pack)
Synonyme(s) :
ThermASeal Foil heat sealable seal, ThermASeal Foil Sealing Film, 78mm x 125mm, 5 sheets, bonding foil, non pierable bonding foil
About This Item
Code UNSPSC :
41122410
Nomenclature NACRES :
NB.15
Stérilité
non-sterile
Fabricant/nom de marque
IST Scientific IST-108-078TS
Conditionnement
pkg of pack of 5 sheets (trial pack)
Technique(s)
PCR: suitable
Larg. × L
78 mm × 125 mm
Adéquation
suitable for PCR
Description générale
Strong bonding foil to polypropylene, ideal for water thermal cyclers.
Features: Foil laminate, strong bonding, good solvent resistance including DMSO, peelable, non-pierceable.
Compatibility: Polypropylene (PP), Polystyrene (PS).
Seal integrity range: -200°C to 110°C.
Application
PCR, Ideal for water bath thermo cyclers, long term compound storage for hazardous materials
Informations légales
LabSheet is a trademark of IST Scientific
ThermASeal Foil is a trademark of IST Scientific