Principaux documents
EPAG7-TT20
Test Tube Rack, for Eppendorf® New Brunswick Innova® 3100 Water Bath Shakers
holds 29 x 20 mm tubes
1 EA
437.00 CHF
437.00 CHF
Check Cart for Availability
Changer de vue
1 EA
437.00 CHF
About This Item
Code UNSPSC :
41122804
Nomenclature NACRES :
NB.73
437.00 CHF
Check Cart for Availability
Matériaux
stainless steel
Caractéristiques
holds 29 x 20 mm tubes
Fabricant/nom de marque
Eppendorf® AG7-TT20 (North America)
Description générale
Test Tube Racks have an adjustable angle and tube height ­support. Made from stainless-steel with inserts to help retain tubes and ­prevent them from spinning in the rack. Finger-tight knobs let you quickly adjust angle, or release the rack from the base plate, for easy transport to the bench.
Informations légales
Eppendorf is a registered trademark of Eppendorf AG
Innova is a registered trademark of Eppendorf, Inc. 
New Brunswick is a trademark of Eppendorf AG