Principaux documents
EP5920757008
Adapter for Eppendorf® Rotor S-4xUniversal-Large
5/15mL conical tubes and plates, rotor S-4xUniversal Large
Synonyme(s) :
Adapter for Eppendorf® S-4x Universal Large Rotor
About This Item
Code UNSPSC :
41103911
Nomenclature NACRES :
NB.15
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Caractéristiques
5/15mL conical tubes and plates
Conditionnement
pack of 2 ea
Fabricant/nom de marque
5920757008
Compatibilité
rotor S-4xUniversal Large
Description générale
Adapter to hold 24 x 5/15mL conical tubes and plates for use with Eppendorf Rotor S-4xUniversal-Large. Supplied as a pack of 2 adapters.
Adapters for various sizes of tubes and bottles for use with Eppendorf S-4x Universal large rotor.
Informations légales
Eppendorf is a registered trademark of Eppendorf AG