Principaux documents
1.17319
Millipore
RapidMicroBio Base Easy Clean
for use with MAS-100® Iso
1 EACH
4 700.00 CHF
4 700.00 CHF
Date d'expédition estimée le28 mai 2025
Demander des informations complémentaires
Changer de vue
1 EACH
4 700.00 CHF
About This Item
Code UNSPSC :
41122409
Nomenclature NACRES :
NA.74
4 700.00 CHF
Date d'expédition estimée le28 mai 2025
Demander des informations complémentaires
Matériaux
stainless steel base
Agence
non-compendial
Caractéristiques
installed
Application(s)
air monitoring
pharmaceutical
Compatibilité
for use with MAS-100® Iso
Description générale
The RapidMicroBio Base Easy Clean, SST without perforated lid is designed for use with all variants of MAS-100 ISO NT® and MAS-100 ISO MH® remote microbial air samplers.
It is compatible with environmental monitoring cassettes of the Growth Direct® System, made from stainless steel and autoclavable.
This product is compatible with a specific RapidMicroBio Perforated Lid designed for Growth Direct® cassettes (article 1.17314).
Application
The RapidMicroBio Base Easy Clean, SST without perforated lid for MAS-100 ISO NT®/ MAS-100 ISO MH® remote microbial air samplers is used for viable air monitoring in isolators and RABS with Growth Direct® environmental monitoring cassettes.
Informations légales
Growth Direct is a registered trademark of Rapid Micro Biosystems, Inc.
MAS-100 is a registered trademark of MBV AG
MAS-100 ISO MH is a registered trademark of MBV AG
MAS-100 ISO NT is a registered trademark of MBV AG