Principaux documents
Z645281
IKA® sieves for MF 10 basic microfine grinder drive
MF 2.0, 2.0mm, 1/cs
1 EA
387.00 CHF
387.00 CHF
Check Cart for Availability
Changer de vue
1 EA
387.00 CHF
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.74
387.00 CHF
Check Cart for Availability
Conditionnement
case of 1
pack of 1
Fabricant/nom de marque
IKA 2939400
Taille
2.0 mm
Description générale
Note: Interchangeable sieve for insertion into the grinding heads ensures maximum particle size filtering.
Informations légales
IKA is a registered trademark of IKA-Werke GmbH & Co. KG