Principaux documents
Z645273
IKA® sieves for MF 10 basic microfine grinder drive
MF 1.0, 1.0mm, 1/cs
1 EA
400.00 CHF
400.00 CHF
Check Cart for Availability
Changer de vue
1 EA
400.00 CHF
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.74
400.00 CHF
Check Cart for Availability
Conditionnement
case of 1
pack of 1
Fabricant/nom de marque
IKA 2939200
Taille
1.0 mm
Description générale
Note: Interchangeable sieve for insertion into the grinding heads ensures maximum particle size filtering.
Informations légales
IKA is a registered trademark of IKA-Werke GmbH & Co. KG
Alexey Veremeev et al.
Materials (Basel, Switzerland), 13(15) (2020-08-06)
Here we evaluated the efficacy of bone repair using various native bovine biomaterials (refined hydroxyapatite (HA), demineralised bone matrix (DBM), and purified bone collagen (COLL)) as compared with commercially available bone mineral and bone autografts. We employed a conventional critical-sized