Principaux documents
Z567582
Sigma-Aldrich
Aldrich® Benchclamp 2-prong
dual adjustment grippy knobs, size L
Synonyme(s) :
bench clamps
1 EA
57.60 CHF
57.60 CHF
Check Cart for Availability
Changer de vue
1 EA
57.60 CHF
About This Item
Code UNSPSC :
41122413
Nomenclature NACRES :
NB.35
57.60 CHF
Check Cart for Availability
Taille
L
Capacité de la mâchoire
0-65 mm
Largeur de la fixation
23 mm
Description générale
Aldrich benchclamp clamps have a nickel-plated finish and are corrosion resistant. The clamps come with slip-on fiberglass finger covers. The Grippy Knob design has a round nut which is easy to turn making tightening the clamps around glassware fast and convenient.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Benchclamp is a trademark of Sigma-Aldrich Co. LLC