Principaux documents
Z562491
LabJaws regular clamp holder
jaw capacity 0-18 mm , 0-71 inches, nickel plated (zinc)
1 EA
31.80 CHF
31.80 CHF
Check Cart for Availability
Changer de vue
1 EA
31.80 CHF
About This Item
Code UNSPSC :
41122413
Nomenclature NACRES :
NB.35
31.80 CHF
Check Cart for Availability
Matériaux
nickel plated (zinc)
Fabricant/nom de marque
Troemner 916051
Capacité de la mâchoire
0-18 mm , 0-71 inches
Description générale
Corrosion resistant. Has precise fit and powerful holding ability. Clamps at a 90 degree angle.
Informations légales
LabJaws is a trademark of Henry Troemner LLC