Principaux documents
Z551899
Sigma-Aldrich
Aldrich® in-line oil bubbler with SafetyBarb® hose connectors
joint: ST/NS 19/22
Synonyme(s) :
bubblers, in-line bubblers, scientific bubblers
1 EA
213.00 CHF
213.00 CHF
Check Cart for Availability
Changer de vue
1 EA
213.00 CHF
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
213.00 CHF
Check Cart for Availability
Joint
joint: ST/NS19/22
Capacité
10 mL
Catégories apparentées
Description générale
For precise N2 pressure control during inert atmosphere reactions. Connect reaction vessel to in-line joint or use with a ballast bulb to keep pressure constant.
Order replacement SafetyBarb hose connectors Z547786 (straight) or Z547883 (angled).
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
SafetyBarb is a registered trademark of Merck KGaA, Darmstadt, Germany