Principaux documents
Z517127
Sigma-Aldrich
Aldrich® 75 mm O-ring top
Torion inlet top
1 EA
359.00 CHF
359.00 CHF
Check Cart for Availability
Changer de vue
1 EA
359.00 CHF
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.43
359.00 CHF
Check Cart for Availability
Matériaux
glass
Stérilité
non-sterile
Caractéristiques
closure type Standard Taper Joint
Conditionnement
pack of 1
Description générale
Each top comes with a 4 mm PTFE stopcock. Use the stainless steel clamp Z517275.
Torion inlet style − Use 1/4 in. hard-wall transfer tubing (e.g. PP or PTFE) for the airtight transfer of air-sensitive materials.
Standard taper − Joint can accommodate a transfer adapter or a filter funnel.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Torion is a trademark of Bibby Sterlin Ltd.
À utiliser avec