Principaux documents
Z503282
Sigma-Aldrich
Scienceware® Delrin® screw clamp
pkg of 3 ea
3 EA
38.20 CHF
38.20 CHF
Check Cart for Availability
Changer de vue
3 EA
38.20 CHF
About This Item
Code UNSPSC :
41122413
Nomenclature NACRES :
NB.35
38.20 CHF
Check Cart for Availability
Conditionnement
pkg of 3 ea
Fabricant/nom de marque
Bel-Art H18212-0000
Description générale
Accepts flexible tubing up to 14 mm O.D. Autoclavable.
Informations légales
Delrin is a registered trademark of E. I. du Pont de Nemours and Company
Scienceware is a registered trademark of SP Scienceware, a division of SP