Principaux documents
Z341894
IKA® dish attachment for KS 130 basic and control shakers
AS 130.3 dish attachment
1 EA
129.00 CHF
129.00 CHF
Date d'expédition estimée le20 mai 2025
Changer de vue
1 EA
129.00 CHF
About This Item
Code UNSPSC :
41121500
Nomenclature NACRES :
NB.73
129.00 CHF
Date d'expédition estimée le20 mai 2025
Matériaux
foil (slip-resistant)
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
IKA 3120000
Description générale
Note: For smooth shaking operations in the low viscosity range, e.g. Petri dishes or culture bottles. With integrated slip-resistant foil (PP).
Informations légales
IKA is a registered trademark of IKA-Werke GmbH & Co. KG