Principaux documents
Z303186
Quickfit® Liebig condenser
L 250 mm, joint: ST/NS 19/26
Synonyme(s) :
C1/12/25/SC
1 EA
59.50 CHF
59.50 CHF
Check Cart for Availability
Changer de vue
1 EA
59.50 CHF
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
59.50 CHF
Check Cart for Availability
Joint
joint: ST/NS19/26
Fabricant/nom de marque
Quickfit C1/12/25/SC
Disponibilité
not available in USA and Canada
not available in USA and Canada
Longueur
250 mm
Superficie
130 cm2
Description générale
Designed to give a turbulent flow in the water jacket for efficient cooling at a comparatively slow rate of water flow. These condensers are adequate and effective in many applications.
Informations légales
Quickfit is a registered trademark of SciLabware Limited