Principaux documents
Z148814
PTFE sealing tape
W 1/4 in.
About This Item
Code UNSPSC :
55121611
Nomenclature NACRES :
NB.35
Largeur
1/4 in.
Taille du rouleau
520 in. (13.2 m)
Description générale
PTFE sealing tape is a chemically inert, self-adhesive tape used in all laboratory plumbing connections, including regulators, control valves, purge valves, in-line flowmeters, impurity traps, gas filters, and ground glass joints. When properly applied to gas or liquid plumbing threads, the PTFE sealing tape fills in gaps between the threads and creates a gas/water-tight seal. When used with ground-glass joints, desiccators, and bell jars, it provides a vacuum-tight seal that is easily removed even after long periods of time.
Application
PTFE sealing tape is commonly used to seal connections in
  • Gas cylinder extraction setups
  • Microfluidic device plumbing setups
  • Carrier gas connections for gas chromatography