Principaux documents
SYNS341750
Synthware high vacuum oblique glass stopcock
bore size 4 mm, plug size 17.35/50
Synonyme(s) :
Synthware S341750
1 EA
114.00 CHF
114.00 CHF
Check Cart for Availability
Changer de vue
1 EA
114.00 CHF
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.51
114.00 CHF
Check Cart for Availability
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware S341750
Taille de l'alésage
4 mm
Catégories apparentées
Description générale
Precision ground with corresponding numbers on the barrel and plug. Lower vacuum chamber is evacuated via a separate 2 mm hole in the plug which is not in the line of flow.
Informations légales
Synthware is a trademark of Kemtech America Inc.