Principaux documents
SYNS721240
Synthware high vacuum T-type stopcock with hollow plug
bore size 2 mm, plug size 12.6/40
Synonyme(s) :
Synthware S721240, T shape stopcock
1 EA
103.00 CHF
103.00 CHF
Check Cart for Availability
Changer de vue
1 EA
103.00 CHF
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.51
103.00 CHF
Check Cart for Availability
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware S721240
Taille de l'alésage
2 mm
Description générale
Precision ground with corresponding numbers on the barrel and plug. Lower vacuum chamber is evacuated via a separate 2 mm hole in the plug which is not in the line of flow.
Informations légales
Synthware is a trademark of Kemtech America Inc.