Principaux documents
SYNA701922
Synthware straight vacuum adapter with PTFE stopcock
male joint: ST/NS 19/22
Synonyme(s) :
Synthware A701922, flow control adapter
1 EA
69.00 CHF
69.00 CHF
Check Cart for Availability
Changer de vue
1 EA
69.00 CHF
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.51
69.00 CHF
Check Cart for Availability
Joint
male joint: ST/NS19/22
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware A701922
Catégories apparentées
Description générale
Used to regulate the flow of gases or liquids into reaction systems. PTFE stopcock has a bore of 2mm. With standard taper inner joint.
Informations légales
Synthware is a trademark of Kemtech America Inc.