Principaux documents
SYNA611922
Synthware straight vacuum adapter with glass stopcock
male joint: ST/NS 19/22
Synonyme(s) :
Synthware A611922, glass adapter with stopcock
1 EA
55.50 CHF
55.50 CHF
Check Cart for Availability
Changer de vue
1 EA
55.50 CHF
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.51
55.50 CHF
Check Cart for Availability
Joint
male joint: ST/NS19/22
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware A611922
Catégories apparentées
Description générale
Used to regulate the flow of gases or liquids into reaction systems. Stopcock has a bore of 2 mm.
Informations légales
Synthware is a trademark of Kemtech America Inc.