Principaux documents
SYNA121922
Synthware vacuum transfer adapter with PTFE stopcocks
joint: ST/NS 19/22
Synonyme(s) :
Synthware A121922, glass vacuum transfer adapter
1 EA
165.00 CHF
165.00 CHF
Check Cart for Availability
Changer de vue
1 EA
165.00 CHF
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.51
165.00 CHF
Check Cart for Availability
Joint
joint: ST/NS19/22
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware A121922
Catégories apparentées
Description générale
With 2 mm PTFE stopcock. For transfer of sensitive and hazardous solutions.
Informations légales
Synthware is a trademark of Kemtech America Inc.