Principaux documents
SYNA121420
Synthware vacuum transfer adapter with PTFE stopcocks
joint: ST/NS 14/20
Synonyme(s) :
Synthware A121420, glass vacuum transfer adapter
1 EA
165.00 CHF
165.00 CHF
Check Cart for Availability
Changer de vue
1 EA
165.00 CHF
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.51
165.00 CHF
Check Cart for Availability
Joint
joint: ST/NS14/20
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware A121420
Catégories apparentées
Description générale
With 2 mm PTFE stopcock. For transfer of sensitive and hazardous solutions.
Informations légales
Synthware is a trademark of Kemtech America Inc.