Principaux documents
DWK73808-28400
Kimble® Closed Top Polypropylene Screw Thread Cap With Welded Ptfe-Faced Silicone Liner
28-400
1 PKG
117.00 CHF
117.00 CHF
Check Cart for Availability
Changer de vue
1 PKG
117.00 CHF
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
NB.17
117.00 CHF
Check Cart for Availability
Matériaux
PTFE/silicone liner
red polypropylene screw cap (Push fit)
Conditionnement
pkg of 1 packs
Fabricant/nom de marque
Kimble® 73808-28400
Raccords
thread
Application
Closure solution for vials and bottles
Caractéristiques et avantages
  • PTFE faced/general purpose white rubber liners are highly resistant to chemical effects
  • Closures are ideal for repeated autoclaving
  • Welded liner technology eliminates the possibility of glue contamination
Informations légales
KIMBLE is a registered trademark of DWK Life Sciences