Principaux documents
CLS4320C50
Pyrex® round-bottom heavy wall boiling flask, short neck with joint
capacity 50 mL, joint: ST/NS 24/40
Synonyme(s) :
bottom flask, flask round, round flask, Corning® 4320C
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Matériaux
borosilicate glass
round bottom flask
short
Joint
joint: ST/NS24/40
Stérilité
non-sterile
Caractéristiques
closure type Standard taper joint
Conditionnement
pack of 1
Fabricant/nom de marque
Corning 4320C-50
Capacité
50 mL
Taille du col
(1)
Description générale
Pyrex heavy duty boiling flask provides increased mechanical strength. The flask features walls that are 25-30% thicker than the standard version.
Informations légales
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.