Principaux documents
CLS43205L
Pyrex® round-bottom boiling flask, short neck
capacity 5,000 mL
Synonyme(s) :
bottom flask, flask glass, round flask
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Matériaux
borosilicate glass
round bottom flask
short
Joint
joint ST/NS45/50
Stérilité
non-sterile
Caractéristiques
closure type Standard taper joint
Conditionnement
case of 4
Fabricant/nom de marque
Corning 4320-5L
Disponibilité
not available in EU
Capacité
5,000 mL
Taille du col
(1)
Description générale
  • PYREX® round-bottom boiling flasks are routinely used in various laboratory applications.
  • Manufactured from low expansion borosilicate glass that provides better clarity, strength, heat, and chemical resistance even in the most demanding laboratory conditions.
  • Precise wall thickness ensures optimal thermal transfer and provides mechanical strength for greater shock resistance.
  • Chemically durable, these flasks are ideal for contamination-free results.
  • The normal working temperature for PYREX® boling flasks is 230 °C.
With full length joints, and short necks.
Informations légales
PYREX is a registered trademark of Corning, Inc.