Principaux documents
BR458733
BRAND® filter crucible
borosilicate glass, capacity 50 mL, 2 D 3
Synonyme(s) :
Filter, Filter crucible
10 EA
381.00 CHF
381.00 CHF
Check Cart for Availability
Changer de vue
10 EA
381.00 CHF
About This Item
Code UNSPSC :
41122200
Nomenclature NACRES :
NB.43
381.00 CHF
Check Cart for Availability
Matériaux
borosilicate glass
Fabricant/nom de marque
BRAND 458733
Capacité
50 mL
Dimension de pores
16-40 μm
Description générale
The BRAND® Filter Crucible made from Boro 3.3 glass is an essential tool for efficient filtration and separation processes in laboratory applications.
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG