Principaux documents
BR148500
BRAND® funnel holder
Holds 1 funnel
Synonyme(s) :
Accessories, Funnels
5 EA
75.20 CHF
75.20 CHF
Check Cart for Availability
Changer de vue
5 EA
75.20 CHF
About This Item
Code UNSPSC :
41121809
Nomenclature NACRES :
NB.35
75.20 CHF
Check Cart for Availability
Matériaux
polypropylene
Caractéristiques
Holds 1 funnel
Conditionnement
pack of 5 ea
Fabricant/nom de marque
BRAND 148500
Description générale
PP. To support 1 or 2 funnels with top outer-Ø of 50 to 120 mm. Easily mounted on supports rods with an outer-Ø 8-14 mm with a locking nut
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG