Principaux documents
BAF371250012
Spinfin® magnetic stirring bar
H × diam. 0.5 in. (12.7 mm) × 0.5 in. (12.7 mm)
Synonyme(s) :
Spinfin® magnetic stirring bar
About This Item
Code UNSPSC :
41122800
Nomenclature NACRES :
NB.51
Matériaux
PTFE-coated
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Bel-Art F37125-0012
H × Diam.
0.5 in. (12.7 mm) × 0.5 in. (12.7 mm)
Description générale
Disks have fins on top for complete agitation.
Spinfin® magnetic stirring bars agitate liquids in round bottom vessels such as test tubes, cylinders and flasks, where other stirring bars cannot be used.
Caractéristiques et avantages
  • Maintains its position in a vessel and will not wobble
  • Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE coating – Superior Chemical Compatibility
  • Manufactured in a registered ISO 9001:2015 facility, as verified by SGS certification
  • Autoclave safe
Informations légales
Spinfin is a registered trademark of SP Scienceware, a division of SP