Skip to Content
Merck
All Photos(1)

Key Documents

EP0030123344

Eppendorf® Safe-Lock micro test tubes

capacity 2.0 mL, PCR clean, colorless, pack of 1000 ea

Synonym(s):

eppendorf tubes, microcentrifuge tubes, microfuge tubes

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
CHF 84.40

CHF 84.40


Check Cart for Availability


Select a Size

Change View
1 PKG
CHF 84.40

About This Item

UNSPSC Code:
41121702
NACRES:
NB.11

CHF 84.40


Check Cart for Availability

material

cap (plug seal)
colorless
colorless cap
colorless tube
round bottom

feature

graduations
neck thread Push fit

packaging

pack of 1000 ea

manufacturer/tradename

Eppendorf® 0030123344

capacity

2.0 mL

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

  • The hinged Eppendorf® Safe-Lock lid provides additional protection against unintentional lid opening during incubation and storage.
  • g-Safe®:Provides great centrifugation stability and prevents sample loss due to tube breakage
  • The optimized sealing properties ensures safety when working with hazardous samples and minimal evaporation rates during long-term storage.
  • The large frosted lid and wide side-surface area make labeling convenient.
  • Functions between temperatures -86 °C to 100 °C
  • Autoclavable when open (121 °C, 20 min)

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
g-Safe is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service