Skip to Content
Merck
All Photos(1)

Documents

EP0030123620

Eppendorf® Safe-Lock microcentrifuge tubes

Forensic DNA Grade, volume 2.0 mL, pack of 500 tubes (10 bags x 50 pcs.)

Synonym(s):

eppendorf tubes, microcentrifuge tubes, microfuge tubes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121703
NACRES:
NB.22

material

cap (Safe-Lock lid)
colorless
polypropylene
polypropylene

sterility

non-sterile

feature

Forensic DNA Grade

packaging

pack of 500 tubes (10 bags x 50 pcs.)

manufacturer/tradename

Eppendorf® 0030123620

parameter

30,000 × g max. RCF

volume

2.0 mL

foreign activity

PCR inhibitors, human DNA, DNase, RNase

PCR inhibitors, human DNA, DNase, RNase, free

Looking for similar products? Visit Product Comparison Guide

General description

Eppendorf® Forensic DNA Grade products comply with the various sophisticated standard requirements of forensic DNA labs that promote automation to reduce the contact between the scientist and the samples, adhering to strict cleaning protocols and procedures, and positive air pressure that blocks the entry of dust particles. It facilitates the prevention of contamination during forensic DNA analysis. This product line is ideal for sample processing, sample storage, DNA extraction, and polymerase chain reaction (PCR) methods

Features and Benefits

  • The hinged safe-lock lid prevents the opening of the lid during storage and incubation
  • Polypropylene provides high resistance to chemicals, high temperatures, and mechanical stress
  • Manufactured without plasticizers, biocides, and slip agents
  • Tight lid sealing prevents evaporation during long-term storage
  • Large, frosted lid and surface on the side for labeling
  • Functions between -86 °C to 100 °C
  • Autoclavable at 121°C for 20 min
  • g-Safe®: Excellent centrifugation stability up to 30,000 × g prevents sample loss due to tube breakage and provides extra safety when working with hazardous samples
Tubes snap closed and stay closed. Flat top with writing surface for positive sample identification

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
g-Safe is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service