Skip to Content
Merck
All Photos(1)

Documents

EP0030120205

Eppendorf® Safe-Lock micro test tubes

tube capacity (2.0 mL), pack of 1000 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.11

material

polypropylene
yellow
yellow cap
yellow tube

packaging

pack of 1000 ea

manufacturer/tradename

Eppendorf® 0030120205

tube capacity

(2.0 mL)

Application

Eppendorf® Safe-Lock micro test tubes are suitable for molecular biology applications including DNA recovery from plasmids.

Features and Benefits

  • The hinged Eppendorf® Safe-Lock lid prevents accidental opening during incubation and storage to keep the sample safe and secure
  • Virgin polypropylene-made tubes have resistance to chemicals, mechanical stress, and high temperature
  • g-Safe®: The Eppendorf Safe-Lock Tubes provide extra safety when handling hazardous samples, with exceptional centrifugation stability up to 30,000 ×g to prevent sample loss due to tube breakage. The level of stability varies depending on the specific tube variant (see manual)
  • Tubes are free of materials that negatively affect bioassays such as mold release agents, softeners, or biocides
  • Precise lid sealing for minimal evaporation rates during long-term storage
  • The large, frosted lid and surface on the side of the tube make it easy to label
  • Ensured functionality from -86 °C to 100 °C
  • Autoclavable at 121 °C for 20 min

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
g-Safe is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service