Merck
Accessibility of N-acyl-D-mannosamines to N-acetyl-D-neuraminic acid aldolase.
Carbohydrate research (2004-07-29)
Yanbin Pan, Tiffany Ayani, Janos Nadas, Shouming Wen, Zhongwu Guo
ZUSAMMENFASSUNG
N-Acetyl-D-neuraminic acid (NeuNAc) aldolase is an important enzyme for the metabolic engineering of cell-surface NeuNAc using chemically modified D-mannosamines. To explore the optimal substrates for this application, eight N-acyl derivatives of D-mannosamine were prepared, and their accessibility to NeuNAc aldolase was quantitatively investigated. The N-propionyl-, N-butanoyl-, N-iso-butanoyl-, N-pivaloyl-, and N-phenylacetyl-D-mannosamines proved to be as good substrates as, or even better than, the natural N-acetyl-D-mannosamine, while the N-trifluoropropionyl and benzoyl derivatives were poor. It was proposed that the electronic effects might have a significant influence on the enzymatic aldol condensation reaction of D-mannosamine derivatives, with electron-deficient acyl groups having a negative impact. The results suggest that N-propionyl-, N-butanoyl-, N-iso-butanoyl-, and N-phenylacetyl-D-mannosamines may be employed to bioengineer NeuNAc on cells.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
3,3,3-Trifluorpropionsäure, 98%